Learn More
Biomatik Corporation Horse Matrix Metalloproteinase 7 (MMP7) CLIA Kit
Supplier:  Biomatik Corporation EKU09595
Encompass_Preferred
Horse Matrix Metalloproteinase 7 (MMP7) CLIA Kit for detecting MMP7 in serum, plasma, other biological fluids. Species Reactivity: Horse (Equus caballus; Equine). Detection Range: 27.4-20,000pg/ml. Sensitivity: 10.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: F6T7U6. Gene ID: 100068985. Target Synonyms: MPSL1; MAT; PUMP-1; Matrilysin Uterine; Matrilysin; Matrin; Pump-1 Protease; Uterine Metalloproteinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5112