Learn More
Biomatik Corporation Horse Matrix Metalloproteinase 1 (MMP1) CLIA Kit
Supplier:  Biomatik Corporation EKU09557
Encompass_Preferred
Horse Matrix Metalloproteinase 1 (MMP1) CLIA Kit for detecting MMP1 in serum, plasma, other biological fluids. Species Reactivity: Horse (Equus caballus; Equine). Detection Range: 13.7-10,000pg/ml. Sensitivity: 5.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9XSZ5. Gene ID: 100033896. Target Synonyms: CLGN; CLG1; Collagenase; Interstitial Collagenase; Vertebrate Collagenase; Fibroblast Collagenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8496