Learn More
American Research Products Inc Horse platelet deriVed growth
Catalog No. 5000327511
Encompass
Horse platelet-deriVed growth factor-BB (PDGF-BB) ELISA kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27511 Supplier American Research Products Inc Supplier No. CSBEQ027241HO
Horse platelet-deriVed growth factor-BB (PDGF-BB) ELISA kit from CUSABIO Detection Range 1 56 ng/mL-100 ng/mL Sensitivity 0 39 ng/mL Sample serum plasma tissue homogenates Method Sandwich