Learn More
Innovative Research Inc Horse RBCs Packed 100 100ml
Catalog No. 502744365
Encompass_Preferred
Horse Red Blood Cells Packed 100% 100ml - Horse Red Blood Cells from Innovative Research are manufactured using whole blood collections from horses. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. The red blood cells are supplied as a 100% suspension in phosphate buffered saline (PBS). Red blood cells are perishable and are collected and processed upon receipt of order.
Catalog No. 50-274-4365 Supplier Innovative Research Inc Supplier No. IHSRBC100P100ML
Horse Red Blood Cells Packed 100% 100ml - Horse Red Blood Cells from Innovative Research are manufactured using whole blood collections from horses. The whole blood is washed to remove the platelet-rich plasma, buffy coat layer, and leukocytes. The red blood cells are supplied as a 100% suspension in phosphate buffered saline (PBS). Red blood cells are perishable and are collected and processed upon receipt of order.