Learn More
Omega Scientific Inc Donor Horse Serum, 500 mL
Catalog No. NC1912620
Encompass_Preferred
Horse Serum contains low trace metals and high protein versus Fetal Bovine Serum. It is collected from controlled donor herds fed a specific diet to maintain consistent serum chemistry. Suitable for use as a mycoplasma growth supplement, in diagnostic assays, and for culturing hematopoietic stem cells and neuronal cells. Virus Screened. Mycoplasma Tested. 0.2 Micron Filtered. 500 mL
Catalog No. NC1912620 Supplier Omega Scientific Inc Supplier No. DH05
Horse Serum contains low trace metals and high protein versus Fetal Bovine Serum. It is collected from controlled donor herds fed a specific diet to maintain consistent serum chemistry. Suitable for use as a mycoplasma growth supplement, in diagnostic assays, and for culturing hematopoietic stem cells and neuronal cells. Virus Screened. Mycoplasma Tested. 0.2 Micron Filtered. 500 mL