Learn More
Sigma Aldrich Fine Chemicals Biosciences Horse Serum Donor herd USA1L
Catalog No. 502904797
Encompass
Horse serum produced from adult horses is homogenous and has low levels of polyamine oxidase. It is metabolically stable and promotes cell proliferation. Horse sera is an alternate to bovine serum and is a source of growth factors and hormones.
Catalog No. 50-290-4797 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H12701L
Horse serum produced from adult horses is homogenous and has low levels of polyamine oxidase. It is metabolically stable and promotes cell proliferation. Horse sera is an alternate to bovine serum and is a source of growth factors and hormones.