Learn More
Omega Scientific Inc Donor Horse Serum, Heat Inactivated, 500 mL
Catalog No. NC1912621
Encompass_Preferred
Horse Serum contains low trace metals and high protein versus Fetal Bovine Serum. It is collected from controlled donor herds fed a specific diet to maintain consistent serum chemistry. Suitable for use as a mycoplasma growth supplement, in diagnostic assays, and for culturing hematopoietic stem cells and neuronal cells. Virus Screened. Mycoplasma Tested. 0.2 Micron Filtered. Product is Heat Inactivated prior to shipment. 500 mL
Catalog No. NC1912621 Supplier Omega Scientific Inc Supplier No. DH07
Horse Serum contains low trace metals and high protein versus Fetal Bovine Serum. It is collected from controlled donor herds fed a specific diet to maintain consistent serum chemistry. Suitable for use as a mycoplasma growth supplement, in diagnostic assays, and for culturing hematopoietic stem cells and neuronal cells. Virus Screened. Mycoplasma Tested. 0.2 Micron Filtered. Product is Heat Inactivated prior to shipment. 500 mL