Learn More
Rockland Immunochemicals Horse Serum Sterile 50 mL
SDP
Catalog No. 501943962
Encompass_Preferred
Horse Serum is used as a supplement to cell culture media. Horse Serum is also suitable for use as a component of bioassays immunoassays or enzyme assays. Horse Serum provides a broad spectrum of macromolecules carrier proteins for lipoid substances and trace elements attachment and spreading factors low molecular weight nutrients and hormones and growth factors that promote cell growth and health. Be certain to maintain Good Cell Culture Practice and maintain sterility of cultures that require media supplementation. Horse Serum is ideal for investigators in Cancer Immunology and Cell Biology research.
Catalog No. 50-194-3962 Supplier Rockland Immunochemicals Supplier No. D107000050
Horse Serum is used as a supplement to cell culture media. Horse Serum is also suitable for use as a component of bioassays immunoassays or enzyme assays. Horse Serum provides a broad spectrum of macromolecules carrier proteins for lipoid substances and trace elements attachment and spreading factors low molecular weight nutrients and hormones and growth factors that promote cell growth and health. Be certain to maintain Good Cell Culture Practice and maintain sterility of cultures that require media supplementation. Horse Serum is ideal for investigators in Cancer Immunology and Cell Biology research.