Learn More
American Research Products Inc Horse Transforming growth fact
Catalog No. 5000327811
Encompass
Horse Transforming growth factor beta-1 (TGFB1) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma Method Competitive
Catalog No. 50-003-27811 Supplier American Research Products Inc Supplier No. CSBEL023446HO
Horse Transforming growth factor beta-1 (TGFB1) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-4000 pg/mL Sensitivity 15 6 pg/mL Sample serum plasma Method Competitive