Learn More
American Research Products Inc Horse tri iodothyronine T3 E
Catalog No. 5000326977
Encompass
Horse tri-iodothyronine (T3) ELISA kit from CUSABIO Detection Range 0 5 ng/mL-8 ng/mL Sensitivity 0 25 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-26977 Supplier American Research Products Inc Supplier No. CSBEQ027510HO
Horse tri-iodothyronine (T3) ELISA kit from CUSABIO Detection Range 0 5 ng/mL-8 ng/mL Sensitivity 0 25 ng/mL Sample serum plasma tissue homogenates Method Competitive