Learn More
American Research Products Inc Horse Tumor necrosis factor a
Catalog No. 5000326752
Encompass
Horse Tumor necrosis factor a TNF-a ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26752 Supplier American Research Products Inc Supplier No. CSBE14031HS
Horse Tumor necrosis factor a TNF-a ELISA Kit from CUSABIO Detection Range 15 6 pg/mL-1000 pg/mL Sensitivity 3 9 pg/mL Sample serum plasma tissue homogenates Method Sandwich