Learn More
Cayman Chemical HorseradIsh PeroxIdase 5ea
SDP
Catalog No. 502892608
Encompass_Preferred
An ionizable cationic lipid (pKa 5.148) LNPs containing 4-O10b1 conjUGated to an anti-F4/80 antibody and encapsulating siRNA targeting the mRNA encoding Tak1 inhibit poly(IC)-induced production of IL-1B IL-6 and TNF-a in RAW 264.7 macrophages intranasal administration of these LNPs decreases lung injury and reduces total protein levels and Mpo activity in BALF in a mouse model of influenza A-induced pneumonia
Catalog No. 50-289-2608 Supplier Cayman Chemical Supplier No. 6000535 EA
An ionizable cationic lipid (pKa 5.148) LNPs containing 4-O10b1 conjUGated to an anti-F4/80 antibody and encapsulating siRNA targeting the mRNA encoding Tak1 inhibit poly(IC)-induced production of IL-1B IL-6 and TNF-a in RAW 264.7 macrophages intranasal administration of these LNPs decreases lung injury and reduces total protein levels and Mpo activity in BALF in a mouse model of influenza A-induced pneumonia