Learn More
Bioworld HORSERADISH PEROXIDASE
Catalog No. 50253324
Encompass_Preferred
Horseradish Peroxidase (HRP) Source:Root extracts of horseradish Description : HRP is widely used as an enzymatic label in immunoassays. Usually, the enzyme is coupled to antibodies, lectins or haptens. Coupling to antibodies etc. ma...
Catalog No. 50-253-324 Supplier Bioworld Supplier No. 220602231
Horseradish Peroxidase (HRP) Source:Root extracts of horseradish Description : HRP is widely used as an enzymatic label in immunoassays. Usually, the enzyme is coupled to antibodies, lectins or haptens. Coupling to antibodies etc. ma...