Learn More
Jackson Immuno Research Labs Peroxidase AffiniPure Bovine Anti-Goat IgG (H+L), .5ml
SDP
Supplier:  Jackson Immuno Research Labs 805035180
Encompass_Preferred
Peroxidase-AffiniPure Bovine Anti-Goat IgG (H+L) (min X Bov,Ck,GP,Sy Hms,Hrs,Hu,Ms,Rb,Rat Sr Prot)
Catalog No. NC9609194