Learn More
World Precision Instrument Horticultural Forceps
SDP
Catalog No. 502539480
Encompass
These forceps are designed for extracting seedling plugs from horiculture trays without damaging the fragile root system.
Catalog No. 50-253-9480 Supplier World Precision Instrument Supplier No. 503411
These forceps are designed for extracting seedling plugs from horiculture trays without damaging the fragile root system.