Learn More
Accuflow Systems Saint-Gobain Food Grade Rubber Hose, 3/4 in I.D., Wire Reinforced
Catalog No. NC3137532
Encompass_Preferred
Saint-Gobain Versilon FGR is a food-grade, wire-reinforced rubber transfer hose engineered for sanitary processing and bulk transfer applications. It combines a food-contact-compatible inner tube with multi-spiral textile and wire reinforcement to deliver flexibility, pressure performance, and wide temperature capability.
  • Food-contact inner tube, FDA and 3-A compliant
  • 0.75 in (19.1 mm) inner diameter, 1.27 in (32.3 mm) outer diameter
  • Maximum working pressure 375 psi (2.59 MPa); minimum burst 1,500 psi (10.34 MPa)
  • Temperature range -40°F to +250°F (-40°C to +121°C)
  • Multiple textile reinforcements with single-helix wire for flexibility
  • Minimum bend radius 2.00 in (50.8 mm); vacuum rating 29.9 in Hg (760 mm Hg)
  • Suitable for sanitary transfer, CIP, washdown, and bulk food handling
Catalog No. NC3137532 Supplier Accuflow Systems Supplier No. HOSFGR12
Saint-Gobain Versilon FGR is a food-grade, wire-reinforced rubber transfer hose engineered for sanitary processing and bulk transfer applications. It combines a food-contact-compatible inner tube with multi-spiral textile and wire reinforcement to deliver flexibility, pressure performance, and wide temperature capability.
  • Food-contact inner tube, FDA and 3-A compliant
  • 0.75 in (19.1 mm) inner diameter, 1.27 in (32.3 mm) outer diameter
  • Maximum working pressure 375 psi (2.59 MPa); minimum burst 1,500 psi (10.34 MPa)
  • Temperature range -40°F to +250°F (-40°C to +121°C)
  • Multiple textile reinforcements with single-helix wire for flexibility
  • Minimum bend radius 2.00 in (50.8 mm); vacuum rating 29.9 in Hg (760 mm Hg)
  • Suitable for sanitary transfer, CIP, washdown, and bulk food handling