Learn More
BURKLE INC PE hose, inner-x outer-Ø 0.24x0.31 in. (6x8 mm), 328.08 in. (100 m)
Catalog No. 5000025790
Encompass_Preferred
The PE hose is characterized by its robustness and reliability. It is suitable for high operating requirements with less flexibility and can be used for pressure applications. It is made of high-quality polyethylene, which gives it excellent resistance to (non-oxidizing) acids, alkalis, salts and a variety of solvents. The hose is very light and dimensionally stable up to + 80 °C. PE hoses have good transparency for optimum visual inspection and have excellent odor and taste neutrality properties.
Catalog No. 50-000-25790 Supplier BURKLE INC Supplier No. 88780608
The PE hose is characterized by its robustness and reliability. It is suitable for high operating requirements with less flexibility and can be used for pressure applications. It is made of high-quality polyethylene, which gives it excellent resistance to (non-oxidizing) acids, alkalis, salts and a variety of solvents. The hose is very light and dimensionally stable up to + 80 °C. PE hoses have good transparency for optimum visual inspection and have excellent odor and taste neutrality properties.