Learn More
Grainger Hose Adapter 1-1/2 NPT 1/4 NPT
Catalog No. 502751623
Encompass_Preferred
The Grainger 1-1/2" NPT Male x 1/4" NPT Female Hose Adapter is made of brass for durability. It allows you to connect a 1-1/2" NPT male threaded pipe to a 1/4" NPT female threaded hose. This adapter is rated for a maximum pressure of 300 PSI and temperature of 250°F. It has a hex shaped body for easy tightening with a wrench. The dimensions are 2-1/2" length x 1-3/4" width. This adapter converts between different thread sizes.
Catalog No. 50-275-1623 Supplier Grainger Supplier No. 14L769
The Grainger 1-1/2" NPT Male x 1/4" NPT Female Hose Adapter is made of brass for durability. It allows you to connect a 1-1/2" NPT male threaded pipe to a 1/4" NPT female threaded hose. This adapter is rated for a maximum pressure of 300 PSI and temperature of 250°F. It has a hex shaped body for easy tightening with a wrench. The dimensions are 2-1/2" length x 1-3/4" width. This adapter converts between different thread sizes.