Learn More
Grainger Hose Adapter 1-1/4 NPTF 1/2 NPTF
Catalog No. 502749797
Encompass_Preferred
The Grainger Approved Hose Adapter is a brass hose adapter that connects 3/4 in. hose ID to 3/4 in. male pipe thread. It has a maximum pressure rating of 300 PSI and temperature rating of 250°F. This adapter allows you to connect a 3/4 in. ID hose to 3/4 in. NPT pipe fittings and valves. It is made of brass for durability and resistance to corrosion. The hex shaped body allows for easy tightening with a wrench.
Catalog No. 50-274-9797 Supplier Grainger Supplier No. 14L776
The Grainger Approved Hose Adapter is a brass hose adapter that connects 3/4 in. hose ID to 3/4 in. male pipe thread. It has a maximum pressure rating of 300 PSI and temperature rating of 250°F. This adapter allows you to connect a 3/4 in. ID hose to 3/4 in. NPT pipe fittings and valves. It is made of brass for durability and resistance to corrosion. The hex shaped body allows for easy tightening with a wrench.