Learn More
Grainger Hose Adapter 1-1/4 NPTF 1/4 NPTF
Catalog No. 502749831
Encompass_Preferred
The Grainger 1-1/4" NPT Female x 1/4" NPT Female Hose Adapter is made from brass for durability. It has a maximum pressure rating of 3000 psi and temperature rating of 250°F. This adapter allows you to connect a 1-1/4" NPT threaded pipe to a 1/4" NPT threaded hose. It can be used in hydraulic, lubrication, refrigeration, instrumentation, industrial, or mobile equipment applications. The dimensions are 2-1/2" length x 1-5/16" width. It weighs 0.19 lbs. This adapter provides a secure threaded connection between mismatched thread sizes.
Catalog No. 50-274-9831 Supplier Grainger Supplier No. 14L774
The Grainger 1-1/4" NPT Female x 1/4" NPT Female Hose Adapter is made from brass for durability. It has a maximum pressure rating of 3000 psi and temperature rating of 250°F. This adapter allows you to connect a 1-1/4" NPT threaded pipe to a 1/4" NPT threaded hose. It can be used in hydraulic, lubrication, refrigeration, instrumentation, industrial, or mobile equipment applications. The dimensions are 2-1/2" length x 1-5/16" width. It weighs 0.19 lbs. This adapter provides a secure threaded connection between mismatched thread sizes.