Learn More
Grainger Hose Adapter 1-1/4 NPTM 3/8 NPTM
Catalog No. 502749830
Encompass_Preferred
The Grainger Approved Hose Adapter is a brass hose adapter that connects 3/4 in. hose ID to 1/2 in. male pipe thread. It has a maximum pressure rating of 300 PSI and temperature rating of 250°F. This adapter allows you to connect a 3/4 in. ID hose to 1/2 in. NPT pipe fittings and valves. Made from brass for durability and corrosion resistance.
Catalog No. 50-274-9830 Supplier Grainger Supplier No. 14L772
The Grainger Approved Hose Adapter is a brass hose adapter that connects 3/4 in. hose ID to 1/2 in. male pipe thread. It has a maximum pressure rating of 300 PSI and temperature rating of 250°F. This adapter allows you to connect a 3/4 in. ID hose to 1/2 in. NPT pipe fittings and valves. Made from brass for durability and corrosion resistance.