Learn More
Grainger Hose Adapter 1/2 JIC 1/2 BSPP
Catalog No. 502763799
Encompass_Preferred
The ADAPTALL Hose Adapter is a 1/2" JIC female to 1/2" BSPP male adapter made of zinc-plated steel. It has a maximum working pressure of 3,000 psi and temperature range of -40°F to 250°F. This adapter allows you to connect hoses with different thread types. It has a hex body for easy tightening and can be used with hydraulic hoses, lubrication systems, brake lines, transmission coolers, power steering systems, and more. The zinc plating resists corrosion. This adapter is made in the USA and conforms to SAE J514 specifications. It comes with a Grainger 5-year warranty.
Catalog No. 50-276-3799 Supplier Grainger Supplier No. 16P912
The ADAPTALL Hose Adapter is a 1/2" JIC female to 1/2" BSPP male adapter made of zinc-plated steel. It has a maximum working pressure of 3,000 psi and temperature range of -40°F to 250°F. This adapter allows you to connect hoses with different thread types. It has a hex body for easy tightening and can be used with hydraulic hoses, lubrication systems, brake lines, transmission coolers, power steering systems, and more. The zinc plating resists corrosion. This adapter is made in the USA and conforms to SAE J514 specifications. It comes with a Grainger 5-year warranty.