Learn More
Grainger Hose Adapter 1/2 NPTF 1-1/2 NPTF
Catalog No. 502764520
Encompass_Preferred
The ADAPTALL Hydraulic Hose Adapter is a 1 inch hydraulic hose adapter made of steel with a zinc finish. It has a working pressure of 5000 psi and temperature range of -40 to 212 degrees Fahrenheit. This adapter allows you to connect hoses with different thread types. It has a JIC 37 degree female thread on one end and a SAE O-ring boss thread on the other end. The adapter is made in the USA and converts between metric and SAE thread types. It can be used with hydraulic hoses, pumps, valves, and cylinders. This adapter provides a high quality solution for connecting mismatched hydraulic components.
Catalog No. 50-276-4520 Supplier Grainger Supplier No. 16P920
The ADAPTALL Hydraulic Hose Adapter is a 1 inch hydraulic hose adapter made of steel with a zinc finish. It has a working pressure of 5000 psi and temperature range of -40 to 212 degrees Fahrenheit. This adapter allows you to connect hoses with different thread types. It has a JIC 37 degree female thread on one end and a SAE O-ring boss thread on the other end. The adapter is made in the USA and converts between metric and SAE thread types. It can be used with hydraulic hoses, pumps, valves, and cylinders. This adapter provides a high quality solution for connecting mismatched hydraulic components.