Learn More
Grainger Hose Adapter 1/2 NPTF 1/2 NPTF
Catalog No. 502764522
Encompass_Preferred
The 1/2" NPTF Female x 1/2" NPTF Female Hose Adapter from Grainger is made of brass for durability. It allows you to connect two male threaded fittings or hoses together. With a pressure rating of up to 3000 PSI, this adapter is suitable for high pressure hydraulic applications. The hex shaped body allows for easy tightening with a wrench. This adapter conforms to ASME B1.20.1 standards. Overall it provides a reliable solution for joining two male threaded hydraulic connections.
Catalog No. 50-276-4522 Supplier Grainger Supplier No. 16P927
The 1/2" NPTF Female x 1/2" NPTF Female Hose Adapter from Grainger is made of brass for durability. It allows you to connect two male threaded fittings or hoses together. With a pressure rating of up to 3000 PSI, this adapter is suitable for high pressure hydraulic applications. The hex shaped body allows for easy tightening with a wrench. This adapter conforms to ASME B1.20.1 standards. Overall it provides a reliable solution for joining two male threaded hydraulic connections.