Learn More
Grainger Hose Adapter 1/2 NPTF 1/2 NPTF
Catalog No. 502764149
Encompass_Preferred
The ADAPTALL Hydraulic Hose Adapter is a 3/4" hydraulic hose adapter made of steel with a zinc finish. It has a working pressure of 5,000 psi and temperature range of -40°F to +212°F. This adapter connects a 3/4" hydraulic hose to 3/4" JIC fittings. It is manufactured in the USA by Adaptall. The adapter body is made of steel for durability with a zinc finish to resist corrosion. It can be used in a variety of hydraulic applications.
Catalog No. 50-276-4149 Supplier Grainger Supplier No. 16P931
The ADAPTALL Hydraulic Hose Adapter is a 3/4" hydraulic hose adapter made of steel with a zinc finish. It has a working pressure of 5,000 psi and temperature range of -40°F to +212°F. This adapter connects a 3/4" hydraulic hose to 3/4" JIC fittings. It is manufactured in the USA by Adaptall. The adapter body is made of steel for durability with a zinc finish to resist corrosion. It can be used in a variety of hydraulic applications.