Learn More
Grainger Hose Adapter 1/2 NPTF 1 NPTF
Catalog No. 502764691
Encompass_Preferred
The 1/2" NPTF Female x 1" NPTF Male Hose Adapter from Grainger is made from brass for durability. It allows you to connect a 1/2" NPTF threaded hose end to a 1" NPTF threaded pipe fitting. This adapter can handle pressures up to 3000 psi and temperatures up to 250°F. It has a hex body for easy tightening with a wrench. The brass construction makes it suitable for air, water, oil, natural gas, and inert gas applications. Overall, this adapter provides a reliable connection between different thread sizes.
Catalog No. 50-276-4691 Supplier Grainger Supplier No. 16P918
The 1/2" NPTF Female x 1" NPTF Male Hose Adapter from Grainger is made from brass for durability. It allows you to connect a 1/2" NPTF threaded hose end to a 1" NPTF threaded pipe fitting. This adapter can handle pressures up to 3000 psi and temperatures up to 250°F. It has a hex body for easy tightening with a wrench. The brass construction makes it suitable for air, water, oil, natural gas, and inert gas applications. Overall, this adapter provides a reliable connection between different thread sizes.