Learn More
Grainger Hose Adapter 1/2 NPTF 1 NPTF
Catalog No. 502763800
Encompass_Preferred
The ADAPTALL Hydraulic Hose Adapter is a 3/4" hydraulic hose adapter made by Adaptall. It is designed to connect hydraulic hoses to hydraulic equipment. This adapter is made from steel and has a maximum working pressure of 5,000 psi. It has a JIC 37° flare on one end to connect to hydraulic hose and 3/4" NPT threads on the other end to connect to hydraulic components. The adapter allows you to connect hydraulic hoses to pumps, valves, cylinders, and motors. It provides a high pressure seal and is reusable. This adapter meets or exceeds SAE J514 specifications.
Catalog No. 50-276-3800 Supplier Grainger Supplier No. 16P933
The ADAPTALL Hydraulic Hose Adapter is a 3/4" hydraulic hose adapter made by Adaptall. It is designed to connect hydraulic hoses to hydraulic equipment. This adapter is made from steel and has a maximum working pressure of 5,000 psi. It has a JIC 37° flare on one end to connect to hydraulic hose and 3/4" NPT threads on the other end to connect to hydraulic components. The adapter allows you to connect hydraulic hoses to pumps, valves, cylinders, and motors. It provides a high pressure seal and is reusable. This adapter meets or exceeds SAE J514 specifications.