Learn More
Grainger Hose Adapter 1/2 NPTF 3/4 NPTF
Supplier:  Grainger 16P932
Encompass_Preferred
The ADAPTALL Hose Adapter is a versatile adapter that allows you to connect different sized hoses and fittings. It is made from nickel plated brass for durability. The adapter has 1-1/4" NPT female threads on one end and 1-1/4" NPT male threads on the other. It can adapt hoses and fittings from 3/4" to 1-1/4" in size. The maximum pressure rating is 300 PSI and maximum temperature is 250°F. This adapter provides flexibility for connecting mismatched hoses and fittings.
Catalog No. 50-276-4548