Learn More
Grainger Hose Adapter 1/2 NPTF 3/4 NPTF
Catalog No. 502764518
Encompass_Preferred
The 1/2" NPTF Female x 3/4" NPTF Male Hose Adapter is made of brass for durability. It allows you to connect a 3/4" NPTF male fitting to a 1/2" NPTF female fitting. This adapter can handle pressures up to 3000 psi and temperatures up to 250°F. It has a hex body for easy tightening with a wrench. The brass construction makes it suitable for air, water, oil, and inert gas applications. This adapter provides a simple solution for mismatching thread sizes.
Catalog No. 50-276-4518 Supplier Grainger Supplier No. 16P917
The 1/2" NPTF Female x 3/4" NPTF Male Hose Adapter is made of brass for durability. It allows you to connect a 3/4" NPTF male fitting to a 1/2" NPTF female fitting. This adapter can handle pressures up to 3000 psi and temperatures up to 250°F. It has a hex body for easy tightening with a wrench. The brass construction makes it suitable for air, water, oil, and inert gas applications. This adapter provides a simple solution for mismatching thread sizes.