Learn More
Grainger Hose Adapter 1/2 NPTF 3/8 NPTF
Catalog No. 502764547
Encompass_Preferred
The 1/2 NPTF x 3/8 NPTF Hose Adapter from Grainger is a brass fitting that allows you to connect two hoses or pipes with different thread sizes. It has a 1/2 inch NPT female thread on one end and a 3/8 inch NPT female thread on the other end. This adapter is made from brass for durability and resistance to corrosion. It can handle pressures up to 3000 psi and temperatures up to 250°F. This hose adapter provides a secure connection between hoses or pipes with a 1/2 inch male NPT thread and those with a 3/8 inch male NPT thread.
Catalog No. 50-276-4547 Supplier Grainger Supplier No. 16P928
The 1/2 NPTF x 3/8 NPTF Hose Adapter from Grainger is a brass fitting that allows you to connect two hoses or pipes with different thread sizes. It has a 1/2 inch NPT female thread on one end and a 3/8 inch NPT female thread on the other end. This adapter is made from brass for durability and resistance to corrosion. It can handle pressures up to 3000 psi and temperatures up to 250°F. This hose adapter provides a secure connection between hoses or pipes with a 1/2 inch male NPT thread and those with a 3/8 inch male NPT thread.