Learn More
Grainger Hose Adapter 1/4 NPT 1-1/2 NPT
Catalog No. 502763945
Encompass_Preferred
The 1/4 NPT to 1-1/2 NPT Hose Adapter from Grainger is made from brass for durability and resistance to corrosion. It features a 1/4 inch NPT male inlet and a 1-1/2 inch NPT female outlet to connect hoses of different sizes. With a maximum pressure rating of 300 PSI at 70°F, this adapter allows you to connect a 1/4 inch hose to a larger 1-1/2 inch hose or pipe fitting. The hex shaped body provides a tight seal and allows for easy tightening with a wrench.
Catalog No. 50-276-3945 Supplier Grainger Supplier No. 16P937
The 1/4 NPT to 1-1/2 NPT Hose Adapter from Grainger is made from brass for durability and resistance to corrosion. It features a 1/4 inch NPT male inlet and a 1-1/2 inch NPT female outlet to connect hoses of different sizes. With a maximum pressure rating of 300 PSI at 70°F, this adapter allows you to connect a 1/4 inch hose to a larger 1-1/2 inch hose or pipe fitting. The hex shaped body provides a tight seal and allows for easy tightening with a wrench.