Learn More
Grainger Hose Adapter 1/4 NPT 1 NPT
Catalog No. 502764211
Encompass_Preferred
The ADAPTALL Hose Adapter is a 1/4" NPT male to 1" NPT female brass hose adapter. It allows you to connect a 1/4" NPT male fitting to a 1" NPT female hose or pipe fitting. This brass adapter is made in the USA and rated for a maximum pressure of 300 PSI. The hex shaped body allows for easy tightening with a wrench. This adapter enables connecting different sized male and female threaded fittings.
Catalog No. 50-276-4211 Supplier Grainger Supplier No. 16P935
The ADAPTALL Hose Adapter is a 1/4" NPT male to 1" NPT female brass hose adapter. It allows you to connect a 1/4" NPT male fitting to a 1" NPT female hose or pipe fitting. This brass adapter is made in the USA and rated for a maximum pressure of 300 PSI. The hex shaped body allows for easy tightening with a wrench. This adapter enables connecting different sized male and female threaded fittings.