Learn More
Grainger Hose Adapter 1/4 NPTF 1-1/4 NPTF
Catalog No. 502763944
Encompass_Preferred
The Grainger Approved Hose Adapter is a brass fitting that converts 1/4" NPT female pipe threads to 1-1/4" NPT female pipe threads. It allows you to connect two hoses or pipes with different size female threaded ends. This adapter is made from brass for durability and resistance to corrosion. It has a maximum pressure rating of 300 PSI and temperature rating of 250°F. The dimensions are 1-1/2" length x 1-1/4" diameter. This adapter provides a secure threaded connection between mismatched hose or pipe sizes.
Catalog No. 50-276-3944 Supplier Grainger Supplier No. 16P936
The Grainger Approved Hose Adapter is a brass fitting that converts 1/4" NPT female pipe threads to 1-1/4" NPT female pipe threads. It allows you to connect two hoses or pipes with different size female threaded ends. This adapter is made from brass for durability and resistance to corrosion. It has a maximum pressure rating of 300 PSI and temperature rating of 250°F. The dimensions are 1-1/2" length x 1-1/4" diameter. This adapter provides a secure threaded connection between mismatched hose or pipe sizes.