Learn More
Grainger Hose Adapter 1/4 NPTF 1/4 NPTF
Catalog No. 502764692
Encompass_Preferred
The Grainger Approved Hose Adapter is a brass hose adapter that connects 3/4 inch hose to 1/2 inch pipe thread. It has a maximum pressure rating of 300 PSI and temperature rating of 250 degrees Fahrenheit. The adapter is made of forged brass for durability and has a hex head for easy tightening. It can be used for air, water, oil, and inert gas applications. The adapter allows you to connect a 3/4 inch ID hose to 1/2 inch pipe thread fittings or valves.
Catalog No. 50-276-4692 Supplier Grainger Supplier No. 16P922
The Grainger Approved Hose Adapter is a brass hose adapter that connects 3/4 inch hose to 1/2 inch pipe thread. It has a maximum pressure rating of 300 PSI and temperature rating of 250 degrees Fahrenheit. The adapter is made of forged brass for durability and has a hex head for easy tightening. It can be used for air, water, oil, and inert gas applications. The adapter allows you to connect a 3/4 inch ID hose to 1/2 inch pipe thread fittings or valves.