Learn More
Grainger Hose Adapter 1/4 NPTF 1/4 NPTF
Catalog No. 502764145
Encompass_Preferred
The ADAPTALL Hydraulic Hose Adapter from Parker is a 5/16" male JIC 37° flare x 1/4" female NPT straight thread adapter fitting. It is made from zinc-plated steel for corrosion resistance and has a maximum working pressure of 5000 psi. This adapter allows you to connect a 1/4" NPT threaded hydraulic component to a 5/16" JIC hydraulic hose. It can be used in a variety of hydraulic applications. The adapter is sold individually.
Catalog No. 50-276-4145 Supplier Grainger Supplier No. 16P907
The ADAPTALL Hydraulic Hose Adapter from Parker is a 5/16" male JIC 37° flare x 1/4" female NPT straight thread adapter fitting. It is made from zinc-plated steel for corrosion resistance and has a maximum working pressure of 5000 psi. This adapter allows you to connect a 1/4" NPT threaded hydraulic component to a 5/16" JIC hydraulic hose. It can be used in a variety of hydraulic applications. The adapter is sold individually.