Learn More
Grainger Hose Adapter 1/4 NPTF 1/4 NPTF
Catalog No. 502764574
Encompass_Preferred
The Grainger Approved Hose Adapter is a 1/4" NPT male x 1/4" barb fitting made of brass. It can be used to connect 1/4" tubing to 1/4" NPT threaded pipe connections. This adapter has a pressure rating of up to 250 PSI and temperature range of -40°F to 250°F. The barbed end accepts 1/4" ID hose or tubing. This fitting allows you to quickly connect flexible tubing in areas where you need to route fluid lines. It provides a secure seal while maintaining flexibility.
Catalog No. 50-276-4574 Supplier Grainger Supplier No. 16P925
The Grainger Approved Hose Adapter is a 1/4" NPT male x 1/4" barb fitting made of brass. It can be used to connect 1/4" tubing to 1/4" NPT threaded pipe connections. This adapter has a pressure rating of up to 250 PSI and temperature range of -40°F to 250°F. The barbed end accepts 1/4" ID hose or tubing. This fitting allows you to quickly connect flexible tubing in areas where you need to route fluid lines. It provides a secure seal while maintaining flexibility.