Learn More
Grainger Hose Adapter 1/4 NPTF 12 mm
Catalog No. 502764151
Encompass_Preferred
The ADAPTALL Hydraulic Hose Adapter is a 1/4" NPTF (female) to 12 mm hydraulic hose adapter made of steel with a zinc finish. It has a maximum working pressure of 5,000 psi and operating temperature range of -40°F to +212°F. This adapter allows you to connect 1/4" NPTF threaded ports to 12 mm hydraulic hose. It is manufactured by Adaptall, comes in a 1-pack, and can be used in hydraulic, lubrication, and industrial applications. The dimensions are 1-1/4" length x 1-1/16" width.
Catalog No. 50-276-4151 Supplier Grainger Supplier No. 16P939
The ADAPTALL Hydraulic Hose Adapter is a 1/4" NPTF (female) to 12 mm hydraulic hose adapter made of steel with a zinc finish. It has a maximum working pressure of 5,000 psi and operating temperature range of -40°F to +212°F. This adapter allows you to connect 1/4" NPTF threaded ports to 12 mm hydraulic hose. It is manufactured by Adaptall, comes in a 1-pack, and can be used in hydraulic, lubrication, and industrial applications. The dimensions are 1-1/4" length x 1-1/16" width.