Learn More
Grainger Hose Adapter 1/4 NPTF 12 mm
Catalog No. 502763802
Encompass_Preferred
The ADAPTALL Hydraulic Hose Adapter is a 3/8" NPT male to 3/8" NPT female hydraulic hose adapter made of steel with a zinc finish. It has a maximum working pressure of 5,000 psi and operating temperature range of -40°F to 250°F. This adapter allows you to connect a 3/8" NPT male fitting to a 3/8" NPT female hydraulic hose fitting. It is manufactured in the USA by Adaptall.
Catalog No. 50-276-3802 Supplier Grainger Supplier No. 16P941
The ADAPTALL Hydraulic Hose Adapter is a 3/8" NPT male to 3/8" NPT female hydraulic hose adapter made of steel with a zinc finish. It has a maximum working pressure of 5,000 psi and operating temperature range of -40°F to 250°F. This adapter allows you to connect a 3/8" NPT male fitting to a 3/8" NPT female hydraulic hose fitting. It is manufactured in the USA by Adaptall.