Learn More
Grainger Hose Adapter 1/4 NPTF 14 mm
Catalog No. 502764714
Encompass_Preferred
The ADAPTALL Hydraulic Hose Adapter is a 1/4" NPT male to 1/4" NPT female hydraulic hose adapter made of steel with a zinc finish. It has a maximum working pressure of 5,000 psi and operating temperature range of -40°F to 250°F. This adapter allows you to connect 1/4" NPT threaded ports to 1/4" hydraulic hose. It is manufactured in the USA by Adaptall.
Catalog No. 50-276-4714 Supplier Grainger Supplier No. 16P942
The ADAPTALL Hydraulic Hose Adapter is a 1/4" NPT male to 1/4" NPT female hydraulic hose adapter made of steel with a zinc finish. It has a maximum working pressure of 5,000 psi and operating temperature range of -40°F to 250°F. This adapter allows you to connect 1/4" NPT threaded ports to 1/4" hydraulic hose. It is manufactured in the USA by Adaptall.