Learn More
Grainger Hose Adapter 1/4 NPTF 3/4 NPTF
Catalog No. 502763801
Encompass_Preferred
The ADAPTALL Hydraulic Hose Adapter is a 3/4" hydraulic hose adapter made by Adaptall. It is designed to connect hoses with different thread types and sizes. This adapter has a 3/4" JIC 37° female swivel nut on one end and a 3/4" SAE O-ring boss on the other end. It is made from zinc plated steel for durability. The operating pressure is up to 5000 psi and the temperature range is from -40°F to 250°F. This adapter allows you to connect hoses with incompatible fittings.
Catalog No. 50-276-3801 Supplier Grainger Supplier No. 16P934
The ADAPTALL Hydraulic Hose Adapter is a 3/4" hydraulic hose adapter made by Adaptall. It is designed to connect hoses with different thread types and sizes. This adapter has a 3/4" JIC 37° female swivel nut on one end and a 3/4" SAE O-ring boss on the other end. It is made from zinc plated steel for durability. The operating pressure is up to 5000 psi and the temperature range is from -40°F to 250°F. This adapter allows you to connect hoses with incompatible fittings.