Learn More
Grainger Hose Adapter 1/8 NPT 22 mm
Catalog No. 502763807
Encompass_Preferred
The ADAPTALL Hydraulic Hose Adapter is a 3/4" hydraulic hose adapter made of steel with a zinc finish. It has a working pressure of 5,000 psi and temperature range of -40°F to +212°F. This adapter allows you to connect a 3/4" hydraulic hose to 3/4" JIC fittings. It is manufactured in the USA by Adaptall. The adapter body is made of steel for durability with a zinc finish to resist corrosion. It can adapt a 3/4" hydraulic hose to connect to 3/4" JIC male and female threaded ports. This versatile adapter enables connecting hydraulic hoses where you need a compact, heavy-duty solution.
Catalog No. 50-276-3807 Supplier Grainger Supplier No. 16P949
The ADAPTALL Hydraulic Hose Adapter is a 3/4" hydraulic hose adapter made of steel with a zinc finish. It has a working pressure of 5,000 psi and temperature range of -40°F to +212°F. This adapter allows you to connect a 3/4" hydraulic hose to 3/4" JIC fittings. It is manufactured in the USA by Adaptall. The adapter body is made of steel for durability with a zinc finish to resist corrosion. It can adapt a 3/4" hydraulic hose to connect to 3/4" JIC male and female threaded ports. This versatile adapter enables connecting hydraulic hoses where you need a compact, heavy-duty solution.