Learn More
Grainger Hose Adapter 1/8 NPT 26 mm
Catalog No. 502764073
Encompass_Preferred
The ADAPTALL Hydraulic Hose Adapter 1/8 NPT Male x 1/8 NPT Female is a hydraulic hose adapter that allows you to connect two hydraulic hoses with 1/8" NPT thread sizes. It is made from zinc-plated steel for durability. This adapter has a maximum working pressure of 5000 PSI and temperature range of -40°F to 250°F. The dimensions are 1-1/2" length x 15/16" width. This hydraulic hose adapter can be used for hydraulic applications to connect hoses where both ends have a 1/8" NPT thread. It provides a secure connection between hydraulic hoses.
Catalog No. 50-276-4073 Supplier Grainger Supplier No. 16P951
The ADAPTALL Hydraulic Hose Adapter 1/8 NPT Male x 1/8 NPT Female is a hydraulic hose adapter that allows you to connect two hydraulic hoses with 1/8" NPT thread sizes. It is made from zinc-plated steel for durability. This adapter has a maximum working pressure of 5000 PSI and temperature range of -40°F to 250°F. The dimensions are 1-1/2" length x 15/16" width. This hydraulic hose adapter can be used for hydraulic applications to connect hoses where both ends have a 1/8" NPT thread. It provides a secure connection between hydraulic hoses.