Learn More
Grainger Hose Adapter 1/8 NPTF 27 mm
Catalog No. 502763902
Encompass_Preferred
The ADAPTALL Hydraulic Hose Adapter is a 1/16" NPT male to 1/16" NPT female hydraulic hose adapter made of steel with a zinc finish. It has a maximum working pressure of 5,000 psi and operating temperature range of -40°F to 250°F. This adapter allows you to connect 1/16" NPT threaded ports to 1/16" hydraulic hose. It can be used in hydraulic applications to adapt hose ends to threaded port connections. The all-steel construction provides durability.
Catalog No. 50-276-3902 Supplier Grainger Supplier No. 16P952
The ADAPTALL Hydraulic Hose Adapter is a 1/16" NPT male to 1/16" NPT female hydraulic hose adapter made of steel with a zinc finish. It has a maximum working pressure of 5,000 psi and operating temperature range of -40°F to 250°F. This adapter allows you to connect 1/16" NPT threaded ports to 1/16" hydraulic hose. It can be used in hydraulic applications to adapt hose ends to threaded port connections. The all-steel construction provides durability.