Learn More
Grainger Hose Adapter 1/8 NPTF 42 mm
Catalog No. 502763852
Encompass_Preferred
The Grainger Approved Hose Adapter is a 1/8" NPTF male to 42mm female brass hose adapter. It has a maximum pressure rating of 3000 psi and temperature rating of 250°F. The adapter is made of brass for durability and resistance to corrosion. It can be used to connect hoses or fittings with different thread sizes. The compact adapter allows easy installation in tight spaces. This adapter is ideal for use in hydraulic, pneumatic, lubrication, fuel and oil applications.
Catalog No. 50-276-3852 Supplier Grainger Supplier No. 16P954
The Grainger Approved Hose Adapter is a 1/8" NPTF male to 42mm female brass hose adapter. It has a maximum pressure rating of 3000 psi and temperature rating of 250°F. The adapter is made of brass for durability and resistance to corrosion. It can be used to connect hoses or fittings with different thread sizes. The compact adapter allows easy installation in tight spaces. This adapter is ideal for use in hydraulic, pneumatic, lubrication, fuel and oil applications.