Learn More
Grainger Hose Adapter 1 NPT 5/16 JIC
Catalog No. 502749795
Encompass_Preferred
The 1" NPT to 5/16" JIC hose adapter from Grainger is made of brass and has a maximum pressure rating of 5000 psi. It allows you to connect a 5/16" JIC hose fitting to a 1" NPT threaded port. The adapter has a hex head for easy tightening and can be used for hydraulic applications. Overall dimensions are 1-1/2" length x 1-1/16" width.
Catalog No. 50-274-9795 Supplier Grainger Supplier No. 14L781
The 1" NPT to 5/16" JIC hose adapter from Grainger is made of brass and has a maximum pressure rating of 5000 psi. It allows you to connect a 5/16" JIC hose fitting to a 1" NPT threaded port. The adapter has a hex head for easy tightening and can be used for hydraulic applications. Overall dimensions are 1-1/2" length x 1-1/16" width.