Learn More
Grainger Hose Adapter 1 NPTF 1-1/4 NPTF
Catalog No. 502749746
Encompass_Preferred
The Grainger 1" NPTF to 1-1/4" NPTF Hose Adapter is made of brass for durability. It allows you to connect a 1" NPTF threaded pipe to a 1-1/4" NPTF hose. This adapter can handle pressures up to 250 PSI and temperatures up to 250°F. With zinc plating to resist corrosion. Measures 1-5/8" long with 1-1/4" hose ID. Made in the USA.
Catalog No. 50-274-9746 Supplier Grainger Supplier No. 14L782
The Grainger 1" NPTF to 1-1/4" NPTF Hose Adapter is made of brass for durability. It allows you to connect a 1" NPTF threaded pipe to a 1-1/4" NPTF hose. This adapter can handle pressures up to 250 PSI and temperatures up to 250°F. With zinc plating to resist corrosion. Measures 1-5/8" long with 1-1/4" hose ID. Made in the USA.