Learn More
Grainger Hose Adapter 2 MNPT 2 JIC
Catalog No. 502749799
Encompass_Preferred
The 2-MNPT x 2-JIC Hose Adapter from Grainger is made of steel and has a maximum pressure rating of 5000 psi. It connects a 2" MNPT male pipe thread to a 2" JIC 37° female swivel thread. This adapter allows you to connect hoses to pipes. It can be used in hydraulic applications. The adapter is zinc plated for corrosion resistance. It meets SAE J514 specifications. The approximate dimensions are 3-1/2" length x 2-5/16" width.
Catalog No. 50-274-9799 Supplier Grainger Supplier No. 14L780
The 2-MNPT x 2-JIC Hose Adapter from Grainger is made of steel and has a maximum pressure rating of 5000 psi. It connects a 2" MNPT male pipe thread to a 2" JIC 37° female swivel thread. This adapter allows you to connect hoses to pipes. It can be used in hydraulic applications. The adapter is zinc plated for corrosion resistance. It meets SAE J514 specifications. The approximate dimensions are 3-1/2" length x 2-5/16" width.