Learn More
Grainger Hose Adapter 2 NPTF 12 mm
Catalog No. 502763950
Encompass_Preferred
The ADAPTALL Hydraulic Hose Adapter is a 3/8" NPT male to 3/8" NPT female hydraulic hose adapter made of steel with a zinc finish. It has a maximum working pressure of 5,000 psi and operating temperature range of -40°F to 250°F. This adapter allows you to connect 3/8" NPT threaded ports to 3/8" hydraulic hose. It is manufactured in the USA by Adaptall.
Catalog No. 50-276-3950 Supplier Grainger Supplier No. 16P959
The ADAPTALL Hydraulic Hose Adapter is a 3/8" NPT male to 3/8" NPT female hydraulic hose adapter made of steel with a zinc finish. It has a maximum working pressure of 5,000 psi and operating temperature range of -40°F to 250°F. This adapter allows you to connect 3/8" NPT threaded ports to 3/8" hydraulic hose. It is manufactured in the USA by Adaptall.