Learn More
Grainger Hose Adapter 3/4 in x 22 mm Fitting Size
Catalog No. 502763983
Encompass_Preferred
The Grainger 16P950 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed and fan cooled with Class F insulation and a C-face endshield for easy mounting. It can provide 0.37 kW of output power. The 16P950 would be a good choice for powering pumps, fans, conveyors, compressors, and other industrial equipment.
Catalog No. 50-276-3983 Supplier Grainger Supplier No. 16P950
The Grainger 16P950 is a 1/2 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed and fan cooled with Class F insulation and a C-face endshield for easy mounting. It can provide 0.37 kW of output power. The 16P950 would be a good choice for powering pumps, fans, conveyors, compressors, and other industrial equipment.