Learn More
Grainger Hose Adapter 3/4 NPTF 16 mm
Catalog No. 502765390
Encompass_Preferred
The Grainger Approved Hose Adapter is a 1/4" NPT male x 1/4" barb fitting made of brass. It can be used to connect 1/4" NPT threaded pipe to a 1/4" inside diameter hose. The maximum pressure rating is 3000 psi and the maximum temperature rating is 250°F. This adapter has a hex body for easy tightening and its made in the USA. The overall dimensions are 15/32" width x 1-1/16" length. This fitting allows you to securely connect a 1/4" hose to 1/4" threaded pipe.
Catalog No. 50-276-5390 Supplier Grainger Supplier No. 16P962
The Grainger Approved Hose Adapter is a 1/4" NPT male x 1/4" barb fitting made of brass. It can be used to connect 1/4" NPT threaded pipe to a 1/4" inside diameter hose. The maximum pressure rating is 3000 psi and the maximum temperature rating is 250°F. This adapter has a hex body for easy tightening and its made in the USA. The overall dimensions are 15/32" width x 1-1/16" length. This fitting allows you to securely connect a 1/4" hose to 1/4" threaded pipe.